Learn More
Decon™ CiDecon™ Disinfectant Wipes
Phenolic-based, hard-surface safe to provide broad spectrum efficacy against bacteria, viruses, and fungi, including HIV-1
Supplier: Decon™ 8510
This item is not returnable.
View return policy
Description
Phenolic-based, hard-surface safe to provide broad spectrum efficacy against bacteria, viruses, and fungi, including HIV-1
- Provide broad spectrum efficacy against bacteria, viruses, and fungi, including HIV-1
Compliance
Meets OSHA's Bloodborne Pathogens Standards and is EPA registered.
Specifications
5.75 ±0.5 | |
0.27500% Glutaraldehyde, 0.01375% o-Phenyl phenol, 0.00275% p-Tertiary Amylphenol | |
6 x 10.5 in. | |
Broad spectrum efficacy against Bacteria, Virus and Fungi including HIV-1 on hard, non porous and inanimate surfaces | |
15.24 cm |
EPA, OSHA | |
Pop-up canister | |
Yes | |
6 in. |