Decon™ CiDehol™ ST Sterile Isopropyl Alcohol and Infrared Hands-Free Dispenser Available on GSA/VA Contract for Federal Government customers only.
Sterile, ready-to-use 70% isopropyl alcohol (IPA) solution designed especially for cleanroom use
Manufacturer: Decon Laboratories, Inc.Supplier Diversity Partner 8308
 View more versions of this product
Catalog No. 04-356-16
  Check Availability
Add to Cart
    Specifications
    Certifications/Compliance OSHA
    Sterile Yes
    For Use With (Application) Professional laboratory and healthcare, that uses CiDehol™ ST Sterile 70% IPA in 32 oz. dispenser bag as a disinfectant cleaner where soap and Water are not available
  • Description & Specifications
Related Products
Product Content Feedback