Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Decon™ CiDehol™ ST Sterile 70% Isopropyl Alcohol Solution made with WFI Available on GSA/VA Contract for Federal Government customers only.

Sterile, ready-to-use 70% isopropyl alcohol (IPA) solution designed especially for cleanroom use. Decon Labs, CiDehol™ ST Sterile 70% Isopropyl Alcohol Solution made with WFI is prepared according to USP specifications and packaged in Class 100 cleanroom.

Manufacturer:  Decon Laboratories, Inc.Supplier Diversity Partner 8312

 View more versions of this product

Catalog No. 04-355-500


  Check Availability
Add to cart

Specifications

Description

  • Made with Water-For-Injection
  • Solution is filtered to 0.2µm and individually double-bagged in an ISO Class 5 (Class 100) environment, then sterilized via gamma irradiation
  • Each lot undergoes sterility and pyrogen testing and meets WFI limits in endotoxin
  • Each case shipped with a lot-specific document, which details QC, irradiation, and testing

Description & Specifications

Specifications

Designed especially for cleanroom use
12 oz.
12 oz., 0.35L
70% Isopropyl Alcohol, 30% Water for Injection
Hands-free dispenser
70% Isopropyl Alcohol
CiDehol™ ST Spray
0.35L
USP, ISO 5, EPA
0.2μm
Liquid
Yes

Safety and Handling

ShelfLife : 2 Year(s)

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit