Learn More
Decon™ LeLoop™ Inoculating Loops

Shop All Decon Labs Products


Description
More economical than platinum or disposable plastic loops
Reusable loops are made of high-quality twisted nichrome wire.
- Loops slide smoothly across agar surfaces without tearing or gouging
- The nichrome wire cools rapidly, saving time
- Loops are available calibrated or noncalibrated
- Calibrated in accordance with ASM/APHA via Evans Blue Test and include calibration certificates
- LeLoop™ loops come without holders, which are available separately
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Disposable | Single-use |
| Sterility | Non-sterile |
| Length (English) | 3 in. |
| Length (Metric) | 7.62 cm |
| Volume | 3.41 μL |
| Diameter (Metric) Inside | 0.29 cm |
| Diameter (English) Inside | 0.11 in. |
| Material | Nichrome |
| Size | Medium |
| Product Type | Inoculating Loop |
| Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.