Learn More
Decon™ WFI (Water-for-Injection) Quality Water
Click to view available options
Quantity:
200L
32 oz.
For non-parenteral use
  • Sterile
  • Manufactured according to USP/cGMP guidelines using validated DI/Reverse Osmosis system
  • Used for in vitro applications such as buffer and media prep, dilution of sterile solutions, and production use for cleaning and rinsing equipment
Compliance
Meets USP specifications for WFI.
Quantity 32 oz.