Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Decorin Mouse anti-Human, Alexa Fluor 700, Clone: 5E8E7, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237333AF700

Catalog No. NBP237333X

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

Decorin
5E8E7
Western Blot, Flow Cytometry, ELISA
Purified
DCN
Purified recombinant fragment of human DCN (AA: 263-324) expressed in E. Coli.
Protein A or G purified
Cancer
Primary
1634
ELISA, Flow Cytometry, Western Blot
Alexa Fluor 700
50mM Sodium Borate with 0.05% Sodium Azide
Bone proteoglycan II, CSCD, decorin, decorin proteoglycan, dermatan sulphate proteoglycans II, DSPG2, PG40, PGII, PG-S2, proteoglycan core protein, SLRR1BPGS2, small leucine-rich protein 1B
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit