Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DECR1 Antibody (3B2), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP259469F
This item is not returnable.
View return policy
Description
DECR1 Monoclonal antibody specifically detects DECR1 in Human samples. It is validated for Flow Cytometry, ELISA, ImmunofluorescenceSpecifications
DECR1 | |
Monoclonal | |
FITC | |
2,4-dienoyl CoA reductase 1, mitochondrial, 4-enoyl-CoA reductase, 4-enoyl-CoA reductase [NADPH], DECR, DECR2,4-dienoyl-CoA reductase [NADPH]2,4-dienoyl-CoA reductase, mitochondrial, EC 1.3.1.34, NADPH, SDR18C1, short chain dehydrogenase/reductase family 18C, member 1 | |
Recombinant human DECR1 (35-335aa) purified from E. coli | |
0.1 mL | |
Cancer, Signal Transduction | |
1666 | |
Store at 4°C in the dark. | |
IgG2b κ |
Flow Cytometry, ELISA, Immunofluorescence | |
3B2 | |
PBS | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction