Learn More
DECR1 Antibody (OTI4G4), DyLight 350, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP270564UV
Catalog No. NBP270564UV
Add to Cart
Description
Description
DECR1 Monoclonal specifically detects DECR1 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
Specifications
DECR1
Monoclonal
DyLight 350
50mM Sodium Borate with 0.05% Sodium Azide
DECR1
Full length recombinant protein of human DECR1 (NP_001350) produced in HEK293T cells.
0.1 mL
Primary
Human
IgG1
Western Blot, Immunohistochemistry
OTI4G4
Western Blot, Immunohistochemistry
2,4-dienoyl CoA reductase 1, mitochondrial, 4-enoyl-CoA reductase, 4-enoyl-CoA reductase [NADPH], DECR, DECR2,4-dienoyl-CoA reductase [NADPH]2,4-dienoyl-CoA reductase, mitochondrial, EC 1.3.1.34, NADPH, SDR18C1, short chain dehydrogenase/reductase family 18C, member 1
Mouse
Affinity Purified
Cancer, Signal Transduction
1666.0
Store at 4°C in the dark.
Recommendations
Recommendations