Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DECR1 Mouse anti-Human, Alexa Fluor™ 647, Clone: OTI4G4, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270564AF647

Catalog No. N270564A647


Add to cart

Description

Description

DECR1 Monoclonal antibody specifically detects DECR1 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications

Specifications

DECR1
OTI4G4
Western Blot, Immunohistochemistry
Affinity Purified, Conjugated
DECR1
Full length recombinant protein of human DECR1 (NP_001350) produced in HEK293T cells.
Immunogen affinity purified
Store at 4°C in the dark.
Monoclonal
Human
Immunohistochemistry, Western Blot
Alexa Fluor 647
50mM Sodium Borate with 0.05% Sodium Azide
2,4-dienoyl CoA reductase 1, mitochondrial, 4-enoyl-CoA reductase, 4-enoyl-CoA reductase [NADPH], DECR, DECR2,4-dienoyl-CoA reductase [NADPH]2,4-dienoyl-CoA reductase, mitochondrial, EC 1.3.1.34, NADPH, SDR18C1, short chain dehydrogenase/reductase family 18C, member 1
Mouse
IgG1
0.1 mL
Primary
1666.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit