Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dectin-1/CLEC7A Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 4 publications
$176.85 - $436.00
Specifications
Antigen | Dectin-1/CLEC7A |
---|---|
Concentration | 0.5 mg/mL |
Dilution | Western Blot 0.5 - 4 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
Dectin-1/CLEC7A Polyclonal specifically detects Dectin-1/CLEC7A in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
Dectin-1/CLEC7A | |
Western Blot 0.5 - 4 ug/mL | |
Polyclonal | |
Rabbit | |
Signal Transduction | |
PBS (pH 7.2), 30% glycerol, 0.5% BSA with 0.01% Thimerosal | |
Beta-glucan receptor, BGR, CLECSF12DC-associated C-type lectin 1, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 12, C-type lectin domain family 7 member A, C-type lectin domain family 7, member A, C-type lectin superfamily member 12, dectin-1, DECTIN1CANDF4, Dendritic cell-associated C-type lectin 1, dendritic cell-associated C-type lectin-1, hDectin-1, lectin-like receptor 1 | |
CLEC7A | |
IgG | |
Affinity Purified | |
27.6 kDa |
0.5 mg/mL | |
Western Blot | |
Unconjugated | |
RUO | |
Human, Mouse, Rat | |
Q9BXN2 | |
64581 | |
Synthetic peptide surrounding amino acid 161 of human Dectin-1. | |
Primary | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title