Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dectin-1/CLEC7A Rat anti-Mouse, Alexa Fluor™ 750, Clone: RM0219-9A44, Novus Biologicals™

Rat Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP212259AF750

Catalog No. NP21229A70


Add to cart

Description

Description

Dectin-1/CLEC7A Monoclonal antibody specifically detects Dectin-1/CLEC7A in Mouse samples. It is validated for Western Blot.
Specifications

Specifications

Dectin-1/CLEC7A
RM0219-9A44
Western Blot
Conjugated, Purified
CLEC7A
Mouse Dectin1
Protein G purified
Signal Transduction
Primary
64581.0
Western Blot
Alexa Fluor™ 750
50 mM sodium borate with 0.05% sodium azide
Beta-glucan receptor, BGR, CLECSF12DC-associated C-type lectin 1, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 12, C-type lectin domain family 7 member A, C-type lectin domain family 7, member A, C-type lectin superfamily member 12, dectin-1, DECTIN1CANDF4, Dendritic cell-associated C-type lectin 1, dendritic cell-associated C-type lectin-1, hDectin-1, lectin-like receptor 1
Rat
IgG2
0.1 mL
Store at 4°C in the dark.
Monoclonal
Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit