Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Dectin-2/CLEC6A Antibody Pair [HRP]

Polyclonal/Monoclonal Antibody Pair

Manufacturer:  Novus Biologicals™ NBP2795585PLATES

 View more versions of this product

Catalog No. NB010156


Add to cart

Description

Description

This solid phase sandwich ELISA utilizes an antibody specific for Mouse Dectin-2/CLEC6A.
Specifications

Specifications

Sandwich ELISA
Antibody Pairs
62.5 pg/mL
93978
RUO
Dectin-2/CLEC6A
5 Plates
62.5-4000 pg/mL
CLEC4N, CLECSF10, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 10, C-type lectin domain family 6 member A, C-type lectin domain family 6, member A, C-type lectin superfamily member 10, DC-associated C-type lectin 2, dectin 2, Dectin-2, DECTIN2, Dendritic cell-associated C-type lectin 2
Conjugated
HRP
This solid phase sandwich ELISA utilizes an antibody specific for Mouse Dectin-2/CLEC6A.
Sandwich ELISA
  • Standard: Each vial contains 75 ng of recombinant mouse Dectin-2/CLEC6A.
  • Capture Antibody: 2 mg/mL of rabbit anti-mouse Dectin-2/CLEC6A monoclonal antibody.
  • Detection Antibody: 0.5 mg/mL rabbit anti-Dectin-2/CLEC6A polyclonal antibody conjugated to horseradish-peroxidase (HRP).
  • Storage is content dependent.
    Rabbit
    Mouse
    CLEC6A
    1. Capture Antibody: Dilute to a working concentration of 1.0 μg/mL in CBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.25 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute standard powder with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 4000 pg/mL is recommended.
    SDS
    Documents
    Product Certifications
    Promotions

    Promotions

    Provide Content Correction

    We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Cancel Submit