Learn More
OriGene Dectin 1 (CLEC7A) (NM_197954) Human Tagged ORF Clone Lentiviral Particle, 200 ul
SDP
Supplier:  OriGene RC222007L3V
Encompass_Preferred
Lenti ORF particles, CLEC7A (Myc-DDK tagged) - Human C-type lectin domain family 7, member A (CLEC7A), transcript variant 6, 200ul, >10^7 TU/mL
Catalog No. NC2945588