Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Decynium 22, Tocris Bioscience™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
50mg
This item is not returnable. View return policy
977-96-8
C23H23IN2
454.355
GMYRVMSXMHEDTL-UHFFFAOYSA-M
decynium 22
71299759
CHEBI:37993
(2Z)-1-ethyl-2-[(1-ethylquinolin-1-ium-2-yl)methylidene]quinoline;iodide
CCN1C(=CC2=[N+](C3=CC=CC=C3C=C2)CC)C=CC4=CC=CC=C41.[I-]
This item is not returnable. View return policy
977-96-8
C23H23IN2
454.355
GMYRVMSXMHEDTL-UHFFFAOYSA-M
decynium 22
71299759
CHEBI:37993
(2Z)-1-ethyl-2-[(1-ethylquinolin-1-ium-2-yl)methylidene]quinoline;iodide
CCN1C(=CC2=[N+](C3=CC=CC=C3C=C2)CC)C=CC4=CC=CC=C41.[I-]

PMAT inhibitor

Inhibitor of the plasma membrane monoamine transporter (PMAT) (Ki = 0.10 μM).

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Quantity 50mg
Formula Weight 454.35
Percent Purity >98%
Chemical Name or Material Decynium 22

Recommended Storage : Store at Room Temperature