Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DEDD Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA578280
Description
Positive Control: mouse leukocyte treated with CFA Predicted Reactivity: Rat (98%), Xenopus laevis (86%), Rhesus Monkey (99%), Bovine (97%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
DEDD encodes a protein that contains a death effector domain (DED). DED is a protein-protein interaction domain shared by adaptors, regulators and executors of the programmed cell death pathway. Overexpression of this gene was shown to induce weak apoptosis. Upon stimulation, this protein was found to translocate from cytoplasm to nucleus and colocalize with UBTF, a basal factor required for RNA polymerase I transcription, in the nucleolus. At least three transcript variants encoding the same protein have been found for this gene.
Specifications
DEDD | |
Polyclonal | |
Unconjugated | |
DEDD | |
CASP8IP1; death effector domain containing; death effector domain-containing; death effector domain-containing protein; Death effector domain-containing testicular molecule; DEDD; DEDD1; Dedpro1; DEFT; FLDED1; FLDED-1; KE05; tumor necrosis factor (ligand) superfamily, member 19 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
21945, 9191 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
O75618, Q9Z1L3 | |
DEDD | |
Recombinant protein encompassing a sequence within the center region of human DEDD. The exact sequence is proprietary. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction