Learn More
Thermo Scientific™ MaxQ™ HP Orbital Shakers Dedicated Platforms with Clamps Available on GSA/VA Contract for Federal Government customers only.
Maximize capacity when working with a single-size vessel by using Thermo Scientific™ MaxQ™ HP Orbital Shakers Dedicated Platforms with Clamps.
Manufacturer:  Thermo Scientific™ 238012
 View more versions of this product
Catalog No. 238012
  Check Availability
Add to cart
Specifications
Description
Maximize capacity when working with a single-size vessel by using Thermo Scientific? MaxQ? HP Orbital Shakers Dedicated Platforms with Clamps.
Description & Specifications
Specifications
25
18 in.
45.7cm
125mL Erlenmeyer Flask
MaxQ™ HP 416/420 Shaker
MaxQ HP 416/420
18 in.
45.7cm
Shaker Platform with Clamps