Learn More
Thermo Scientific™ MaxQ™ HP Orbital Shakers Dedicated Platforms with Clamps Available on GSA/VA Contract for Federal Government customers only.
Maximize capacity when working with a single-size vessel by using Thermo Scientific™ MaxQ™ HP Orbital Shakers Dedicated Platforms with Clamps.
Manufacturer:  Thermo Scientific™ 238055
 View more versions of this product
Catalog No. 238055
  Check Availability
Add to cart
Specifications
Description
Maximize capacity when working with a single-size vessel by using Thermo Scientific? MaxQ? HP Orbital Shakers Dedicated Platforms with Clamps.
Description & Specifications
Specifications
4
24 in.
61cm
5L Erlenmeyer Flask
MaxQ™ HP 416 Shaker; Max. 400rpm
MaxQ HP 416; Max. 400rpm
18 in.
45.7cm
Shaker Platform with Clamps