Learn More
Thermo Scientific™ MaxQ™ HP Orbital Shakers Dedicated Platforms with Clamps Available on GSA/VA Contract for Federal Government customers only.
Maximize capacity when working with a single-size vessel by using Thermo Scientific™ MaxQ™ HP Orbital Shakers Dedicated Platforms with Clamps.
Manufacturer:  Thermo Scientific™ 238028
 View more versions of this product
Catalog No. 238028
  Check Availability
Add to cart
Specifications
Description
Maximize capacity when working with a single-size vessel by using Thermo Scientific? MaxQ? HP Orbital Shakers Dedicated Platforms with Clamps.
Description & Specifications
Specifications
70
36 in.
91.4cm
250/300mL Erlenmeyer Flask
MaxQ™ HP 430 Shaker
MaxQ HP 430
24 in.
61cm
Shaker Platform with Clamps