Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Thermo Scientific™ Dedicated Platforms for MaxQ™ 2000/2506/2508/4000/4450/6000 Shakers
Pick the optimal choice for a busy lab, Thermo Scientific™ MaxQ™ dedicated platforms, which are model-specific and come with pre-installed clamps to maximize capacity.
Manufacturer:  Thermo Scientific™ 35247
 View more versions of this product
Catalog No. 11-675-428
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Pick the optimal choice for a busy lab with Thermo Scientific™ Dedicated Platforms for MaxQ™ 2000/2506/2508/4000/4450/6000 Shakers. MaxQ platforms are model-specific and come with pre-installed clamps to maximize capacity. MaxQ Shakers, Platforms and accessories provide exceptional innovation, reliability and accuracy.
Clamps come installed to provide maximum capacity.
Specifications
Specifications
MaxQ 2000, 2506, 2508, 4000, 6000
18 in.
45.7cm
500mL Erlenmeyer flasks
500mL Erlenmeyer flasks
MaxQ™ 2000, 2506, 2508, 4000, and 6000 Shaker
18 in.
45.7cm
16
Erlenmeyer Flask
18 x 18 in. (45.7 x 45.7cm)