Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
dedicator of cytokinesis 8 Antibody, DyLight 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP282084UV
This item is not returnable.
View return policy
Description
dedicator of cytokinesis 8 Polyclonal antibody specifically detects dedicator of cytokinesis 8 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
dedicator of cytokinesis 8 | |
Polyclonal | |
50mM Sodium Borate | |
Rabbit | |
Peptide affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DyLight 350 | |
dedicator of cytokinesis 8, FLJ00026, FLJ00346 | |
Dedicator of cytokinesis 8 antibody was raised against a 17 amino acid peptide near the carboxy terminus of human dedicator of cytokinesis 8. The immunogen is located within the last 50 amino acids of dedicator of cytokinesis 8. Amino Acid Squence: RVESQKRDSFHRSSFRK | |
0.1 mL | |
Cardiovascular Biology, Signal Transduction | |
81704 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction