Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
dedicator of cytokinesis 8 Antibody (OTI4F3), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP246469
Description
Ensure accurate, reproducible results in Immunohistochemistry
dedicator of cytokinesis 8 Monoclonal specifically detects dedicator of cytokinesis 8 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
dedicator of cytokinesis 8 | |
Monoclonal | |
1 mg/mL | |
Immunohistochemistry 1:150 | |
dedicator of cytokinesis 8, FLJ00026, FLJ00346 | |
Mouse | |
238.3 kDa | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
Purified |
Immunohistochemistry | |
OTI4F3 | |
Unconjugated | |
PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide | |
DOCK8 | |
Full length human recombinant protein of human DOCK8(NP_982272) produced in HEK293T cell. | |
Immunogen affinity purified | |
RUO | |
81704 | |
Store at -20C. Avoid freeze-thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction