Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DIEXF Rabbit anti-Hamster, Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548283

Catalog No. PA548283

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Predicted to react with rat based on sequence homology.

Regulates the p53 pathway to control the expansion growth of digestive organs.
Specifications

Specifications

DIEXF
Polyclonal
PBS with 0.09% sodium azide
Q68CQ4, Q8BTT6
Digestive organ expansion factor homolog, DIEXF, C1orf107, DEF
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
100765405, 215193, 27042
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
DIEXF
Liquid
DIEXF
KLH conjugated synthetic peptide between 2-30 amino acids from the N-terminal region of human DEF.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Hamster, Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit