Learn More
Abnova™ DEFA5 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Manufacturer:  Abnova™ H00001670T02
Catalog No. 89-941-752
Add to cart
Specifications
Specifications
Western Blot
293T
NM_021010.1
pCMV-DEFA5 full-length
100 uL
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Human
DEFA5 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 10.1kDa
Transient overexpression cell lysate was tested with anti-DEFA5 antibody (Mfr. No. H00001670-D01P) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)