Learn More
DEFA6 Rabbit anti-Human, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 179231AP
Catalog No. 50-173-1876
Add to cart
Description
Description
Defensins are a family of microbicidal and cytotoxic peptides thought to be involved in host defense. They are abundant in the granules of neutrophils and also found in the epithelia of mucosal surfaces such as those of the intestine, respiratory tract, urinary tract, and vagina. Members of the defensin family are highly similar in protein sequence and distinguished by a conserved cysteine motif. Several alpha defensin genes appear to be clustered on chromosome 8.
Specifications
Specifications
DEFA6
0.27 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q01524
DEF6, DEFA6, Defensin 6, Defensin, alpha 6, defensin, alpha 6, Paneth cell, HD 6
Rabbit
IgG
150 μL
-20°C
Primary
1671
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
DEFA6
Liquid
DEFA6
DEFA6 Fusion Protein Ag12341
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human