Learn More
Azzota Corp Defactinib DFS Item
Manufacturer:  Azzota Corp C1256
PF-04554878 (Defactinib) is an orally bioavailable, small-molecule focal adhesion kinase (FAK) inhibitor with potential antiangiogenic and antineoplastic activities. Please request if need more quantities
Catalog No. 50-161-3163