Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DEFB104A Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | DEFB104A |
---|---|
Dilution | Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:20 - 1:50 |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
DEFB104A Polyclonal specifically detects DEFB104A in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
DEFB104A | |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Human | |
140596 | |
This antibody was developed against a recombinant protein corresponding to the amino acid sequence:EFELDRICGYGTARCRKKCRSQEYRIGRCPNTYACCLRKWDESLLNRT | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
Polyclonal | |
Purified | |
RUO | |
DEFB104B, DEFB-4, defensin, beta 104A, defensin, beta 4 | |
DEFB104A | |
IgG | |
Protein A purified |
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title