Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DEFB107A Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549353

Catalog No. PA549353

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Has antibacterial activity.
Specifications

Specifications

DEFB107A
Polyclonal
PBS with 0.09% sodium azide
Q8IZN7
Beta-defensin 107, Beta-defensin 7, BD-7, DEFB-7, Defensin, beta 107, DEFB107A, DEFB107, DEFB7
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
245910
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
DEFB107A
Liquid
DEFB107A
KLH conjugated synthetic peptide between 56-90 amino acids from the C-terminal region of human DEFB107A.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit