Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DEFB129 Polyclonal Antibody
DEFB129 Polyclonal Antibody
Supplier: Thermo Scientific PA598846
Description
DEFB129 Polyclonal Antibody for Western Blot, IHC (P), ELISA

Specifications
DEFB129 | |
Polyclonal | |
Unconjugated | |
DEFB129 | |
bA530N10.3; BD129; beta defensin 29; beta-defensin 129; Beta-defensin 29; C20orf87; CBD129; DEFB129; DEFB29; DEFB-29; defensin beta 129; defensin, beta 129; hBD-29; prepro-beta-defensin 129 protein; UNQ5794/PRO19599 | |
Rabbit | |
Protein G | |
RUO | |
140881 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
4 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
Q9H1M3 | |
DEFB129 | |
Recombinant Human Beta-defensin 129 protein (20-183AA). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction