Learn More
Ohaus™ DEFENDER™ 3000 Field Test
OHAUS™ DEFENDER™ 3000 Field Test is designed for precision material testing in the construction industry.
Manufacturer:  Ohaus™ 30448500
Catalog No. 01-922-360
Add to cart
Description
Description
  • 125% overload protection
  • Selectable span calibration points
  • Auto-zero tracking
  • Battery-saving auto-off
  • Geo Code geographical gravity calibration adjustment
  • Software menu lockouts
Specifications
Specifications
Digital Field Test Scale
3 sec.
7.4 kg
75 kg
0.02 kg
6-digit, 7-segment LCD with High-contrast White LED Backlight
Product Safety: CAN/CSA C22.2 60950-1, UL 60950-1, IEC/EN 61010-1; Electromagnetic Compatibility: Canada ICES-003 Class B, FCC Part 15 Class B, IEC/EN 61326-1 Class B, Basic environments; Compliance Marks: CE, cULus, RCM
365 x 210 x 400 mm