Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ alpha-Defensin 3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5103123
Description
Antibody detects endogenous levels of total Defensin alpha3.
Defensins are a family of antimicrobial and cytotoxic peptides thought to be involved in host defense. They are abundant in the granules of neutrophils and also found in the epithelia of mucosal surfaces such as those of the intestine, respiratory tract, urinary tract, and vagina. Members of the defensin family are highly similar in protein sequence and distinguished by a conserved cysteine motif. The protein encoded by this gene, defensin, alpha 3, is found in the microbicidal granules of neutrophils and likely plays a role in phagocyte-mediated host defense. Several alpha defensin genes are clustered on chromosome 8. This gene differs from defensin, alpha 1 by only one amino acid.
Specifications
alpha-Defensin 3 | |
Polyclonal | |
Unconjugated | |
DEFA3 | |
alpha-defensin 1; Alpha-defensin 3; cryptdin-1; DEF1; DEF3; Defa1; DEFA1B; DEFA2; Defa27; Defa3; Defcr; Defcr1; Defcr3; Defcr-3; defensin 3, neutrophil-specific; defensin alpha 1; defensin alpha 3; defensin related cryptdin peptide 1; defensin, alpha 1; defensin, alpha 1, myeloid-related sequence; defensin, alpha 2; Defensin, alpha 3; defensin, alpha 3, neutrophil-specific; defensin, alpha, 27; defensin, alpha, 3; defensin-1; defensin-related cryptdin peptide; Defensin-related cryptdin-3; HNP-1; HNP-2; HNP3; HNP-3; HP 1-56; HP 3-56; HP1; HP-1; HP-2; HP3; HP-3; MRS; myeloid-related sequence; neutrophil defensin 1; Neutrophil defensin 2; Neutrophil defensin 3; neutrophil peptide 3 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
13237, 1668 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P28310, P59666 | |
DEFA3 | |
A synthesized peptide derived from human DEFA3(Accession P59666), corresponding to amino acid residues I36-Q86. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction