Learn More
Selleck Chemical LLCSupplier Diversity Partner Deferasirox 10mg 201530-41-8 CGP-72670, ICL-670  DFS Item
Manufacturer:  Selleck Chemical LLCSupplier Diversity Partner S171210MG
Deferasirox (CGP-72670, ICL-670) is an iron chelator, also a cytochrome P450 3A4 inducer, Cytochrome P450 2C8 inhibitor, and Cytochrome P450 1A2 inhibitor. Deferasirox-induced iron depletion promotes BclxL downregulation and cell death.
Catalog No. 50-640-1