Learn More
Medchemexpress LLC HY-13609 50mg , Deflazacort CAS:14484-47-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1360950MG
Encompass_Preferred
HY-13609 50mg Deflazacort CAS: 14484-47-0 Deflazacort, a glucocorticoid, is an inactive prodrug and is converted rapidly to the active metabolite 21-desacetyldeflazacort. Deflazacort is used as an anti-inflammatory and immunosuppressant. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1784
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.