Learn More
Eisco™ Degem™ Electronics Training Modules
SCI_ED
Module topic boards for use with the Degem Educational Electronics Workstation.
$640.00 - $730.00
Specifications
For Use With (Equipment) Degem™ educational electronics workstation
Includes Full course work curriculum
View More Specs
Includes: Full course work curriculum
Requires: Degem Electronics Workstation (S07837), sold separately
Products 2
Catalog Number Mfr. No. Type For Use With (Application) Price Quantity & Availability  
Catalog Number Mfr. No. Type For Use With (Application) Price Quantity & Availability  
S07840
SCI_ED
View Documents
Eisco™
EB3124
Magnetism and Induction Board Training Module Teaches the fundamental concepts of magnetism, electromagnets and transfer including: magnetism, electromagnetism, induction and transformers, essential theory, electromagnetic devices, transformers and troubleshooting exercises
Each for $640.00
Add to cart
S07843
SCI_ED
View Documents
Eisco™
EB3170
Can-Bus Board Training Module Designed to teach the fundamental concepts of the CAN-BUS system and related circuits including: principles of digital systems, CAN-BUS communications, CAN-BUS signals, logic gates and troubleshooting exercises
Each for $730.00
Add to cart
Description
Description
  • Fault insertion mode allows instructors to create an error on the board for students to troubleshoot and develop diagnostic skills
  • Each board requires a minimal amount of wiring to setup
DC Circuits I Board — Teaches the fundamental concepts of DC circuits including: resistors and Ohm's law, voltage sources, resistors in series and Kirchoff's first law, resistors in parallel and Kirchoff's second law, variable resistors and troubleshooting exercises.
AC Circuits Board — Covers the fundamental concepts of AC circuits, signals and filters including: AC circuits, signals and filters, resistors in AC circuits, RC circuits, RL circuits, RLC circuits and troubleshooting exercises.
Magnetism and Induction Board — Teaches the fundamental concepts of magnetism, electromagnets and transfer including: magnetism, electromagnetism, induction and transformers, essential theory, electromagnetic devices, transformers and troubleshooting exercises.
Electrical Control Circuits Board — Allows students to learn the fundamental concepts of electrical components and circuits including: switches, relays, motor and control circuits, temperature control and troubleshooting exercises.
Optical Communication Board — Instructs students in fundamental concepts of digital optical communications including: audio amplifiers and speakers, preamplifiers and speakers, optical transmitters, optical receivers, Schmitt trigger comparators, infrared communications, optical digital communications and troubleshooting exercises.
CAN-BUS Board — Designed to teach the fundamental concepts of the CAN-BUS system and related circuits including: principles of digital systems, CAN-BUS communications, CAN-BUS signals, logic gates and troubleshooting exercises.
Specifications
Specifications
Degem™ educational electronics workstation
Full course work curriculum