Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dehydrated Alcohol, 200 Proof, BiotechGrade, 99.5%, Spectrum™ Chemical
SDP

Catalog Number p-7157391
Click to view available options
Quantity:
500 mL
4 L
200 L
Packaging:
Amber Glass Bottle
Glass Bottle
64-17-5
C2H6O
46.07
LFQSCWFLJHTTHZ-UHFFFAOYSA-N
ethanol
CCO
64-17-5
C2H6O
46.07
LFQSCWFLJHTTHZ-UHFFFAOYSA-N
ethanol
CCO

E1424, 64-17-5, C2H5OH

Spectrum™ Chemical Dehydrated Alcohol, 200 Proof, BiotechGrade The GR is an EMD trademark and indicates this chemical reagent is suitable for use in analytical chemistry and that the product meets or exceeds American Chemical Society (ACS) requirements.
CAS Max % 100%
UN Number UN1170
Percent Purity 99.5%
Grade Biotech
Quantity 500 mL
Packaging Amber Glass Bottle
Water 0.2%
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit