Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dehydrated Alcohol, 200 Proof, Undenatured, USP, 99.5%, Spectrum™

Click to view available options
Quantity:
1 L
100 mL
4 L
4 x 4 L
500 mL
55 gal.
6 x 1 L
6 x 500 mL
Packaging:
Amber Glass Bottle
Drum
64-17-5
C2H6O
46.07
LFQSCWFLJHTTHZ-UHFFFAOYSA-N
ethanol
CCO
64-17-5
C2H6O
46.07
LFQSCWFLJHTTHZ-UHFFFAOYSA-N
ethanol
CCO

ET107, 64-17-5, C2H5OH

Spectrum™ Dehydrated Alcohol, 200 Proof, Undenatured, USP is used as an antiseptic to disinfect skin prior to injections, an active ingredient for pain relief and as a solvent. Spectrum manufacturers Dehydrated Alcohol, 200 Proof, Undenatured, USP and packages it in sizes that various sizes from convenient laboratory 100 mL bottles to 200 L production drums. Like all Spectrum Chemical USP products, dehydrated alcohol products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 100%
UN Number UN1170
Benzene 2 ppm
Percent Purity 99.5%
Methanol 200 ppm
Packaging Amber Glass Bottle
Specific Gravity 0.7962
Quantity 4 L