Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dehydrocholic Acid, USP, 98.5-101%, Spectrum™

Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
5 kg
Packaging:
Poly Bottle
Poly Pail
81-23-2
C24H34O5
402.53
OHXPGWPVLFPUSM-KLRNGDHRSA-N
(4R)-4-[(1R,3aS,3bR,5aS,9aS,9bS,11aR)-9a,11a-dimethyl-4,7,11-trioxo-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl]pentanoic acid
C[C@H](CCC(O)=O)[C@H]1CC[C@H]2[C@H]3[C@H](CC(=O)[C@]12C)[C@@]1(C)CCC(=O)C[C@H]1CC3=O
81-23-2
C24H34O5
402.53
OHXPGWPVLFPUSM-KLRNGDHRSA-N
(4R)-4-[(1R,3aS,3bR,5aS,9aS,9bS,11aR)-9a,11a-dimethyl-4,7,11-trioxo-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl]pentanoic acid
C[C@H](CCC(O)=O)[C@H]1CC[C@H]2[C@H]3[C@H](CC(=O)[C@]12C)[C@@]1(C)CCC(=O)C[C@H]1CC3=O

D1001, 81-23-2, C24H34O5

Spectrum™ Dehydrocholic Acid, USP is used for the treatment of Biliary-tract disorders. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Poly Bottle
Ignition Residue 0.003
Loss on Drying 0.01
Quantity 100 g
Specific Rotation +29.0 to +32.5°
Melting Range 231-242°C
CAS Max % 1
Percent Purity 98.5 to 101%