Learn More
Tocris Bioscience™ DEL 22379
ERK dimerization inhibitor
$203.00 - $840.50
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
57-741-0 View Documents Tocris Bioscience™
5774/10
10mg Each for $203.00
Add to cart
57-745-0 View Documents Tocris Bioscience™
5774/50
Each for $840.50
Add to cart
Description
Description
ERK dimerization inhibitor (IC50 ~ 0.5 μM); inhibits ERK dimerization without affecting phosphorylation or kinase activity. Suppresses growth of tumor cells expressing RAS-ERK oncogenes in vitro. Induces apoptosis and prevents tumor progression in vivo.
Safety and Handling
Safety and Handling
Recommended Storage : Store at −20°C