Delavirdine mesylate, Tocris™

Non-nucleoside reverse transcriptase inhibitor

Manufacturer: R&D Systems 4149/10

Quantity 10mg

 View more versions of this product

Catalog No. 41-491-0

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    Assay Percent Range >99%
    Molecular Formula C22H28N6O3S·CH3SO3H
    Formula Weight 552.67
    Synonym 1-[3-[(1-Methylethyl)amino]-2-pyridinyl]-4-[[5-[(methylsulfonyl)amino]-1H-indol-2-yl]carbonyl]-piperazine monomethanesulfonate
    Chemical Name or Material Delavirdine mesylate

    Specifications

    Formula Weight 552.67
    Quantity 10mg
    Assay Percent Range >99%
    Molecular Formula C22H28N6O3S·CH3SO3H
    CAS 147221-93-0
    Synonym 1-[3-[(1-Methylethyl)amino]-2-pyridinyl]-4-[[5-[(methylsulfonyl)amino]-1H-indol-2-yl]carbonyl]-piperazine monomethanesulfonate
    Recommended Storage Desiccate at Room Temperature
    Chemical Name or Material Delavirdine mesylate

    Desiccate at Room Temperature

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit