Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DELE Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA5144041

Description
Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL.
DELE is a recently identified DAP3-binding protein that is thought to be important in the induction of death receptor-mediated apoptosis. Transfected cells that stably express DELE were found to be susceptible to apoptosis induction by TNF-Alpha and TRAIL, whereas reducing DELE expression by siRNA rescued these cells from apoptosis induction. Furthermore, the reduction of DELE expression also inhibited the activation of caspase-3, caspase-8 and caspase-9 following stimulation by TNF-Alpha, anti-Fas, or TRAIL, indicating the importance of DELE in apoptosis mediated by death receptors.
Specifications
DELE | |
Polyclonal | |
Unconjugated | |
Dele1 | |
0610009O20Rik; 2700004E22Rik; DAP3-binding cell death enhancer 1; Death ligand signal enhancer; Dele; Dele1; KIAA0141; RIKEN cDNA 0610009O20 gene | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9812 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
IgG |
ELISA, Flow Cytometry, Immunocytochemistry, Western Blot | |
500 μg/mL | |
PBS with 4MG trehalose and 0.05MG sodium azide | |
Q14154 | |
Dele1 | |
E.coli-derived human KIAA0141/DELE1 recombinant protein (Position: E218-L430). | |
100 μg | |
Primary | |
Human | |
Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction