Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Delphinidin chloride, ≥95.0% (HPLC), MilliporeSigma™ Supelco™
Delphinidin chloride is commonly found in delphinium flowers. It can be obtained, via hydrolysis of delphinin chloride.
$206.00 - $1493.00
Specifications
CAS | 528-53-0 |
---|---|
Type | Analytical Standard |
Linear Formula | C15H11 ClO7 |
MDL Number | MFCD00016663 |
Synonym | 3,3 ′,4 ′,5,5 ′,7-Hexahydroxyflavylium chloride; 3,5,7-Trihydroxy-2-(3,4,5-trihydroxyphenyl)-1-benzopyrylium chloride |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
11-101-4470
|
MilliporeSigma Supelco
437251MGF |
1 mg |
Each for $206.00
|
|
|||||
11-101-4469
|
MilliporeSigma Supelco
4372510MGF |
10 mg |
Each for $1,493.00
|
|
|||||
Description
Food and BeveragesSpecifications
528-53-0 | |
C15H11 ClO7 | |
3,3 ′,4 ′,5,5 ′,7-Hexahydroxyflavylium chloride; 3,5,7-Trihydroxy-2-(3,4,5-trihydroxyphenyl)-1-benzopyrylium chloride | |
≥95.0% (HPLC) | |
Neat |
Analytical Standard | |
MFCD00016663 | |
338.70 | |
Analytical Standard |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title