Learn More
Delta-Like Protein 1 Mouse anti-Human, BV510, Clone: MHD1-314, BD OptiBuild™
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 744983
Catalog No. BDB744983
Add to cart
Description
Description
Delta-Like Protein 1 Monoclonal antibody specifically detects Delta-Like Protein 1 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
Delta-Like Protein 1
MHD1-314
Brilliant Violet 510
Mouse
50 μg
Refrigerate (2 to 8°C)
Monoclonal
Flow Cytometry
0.2 mg/mL
Purified
IgG1, κ
RUO
Primary
Human