Learn More
Delta Opioid Receptor 358-372 Rabbit anti-Mouse, Rat, Polyclonal, Neuromics™
Rabbit Polyclonal Antibody
Manufacturer:  Neuromics, IncSupplier Diversity Partner RA1010150UL
Encompass
 View more versions of this product
Catalog No. 50-210-0021
Add to cart
Description
Description
Delta Opioid Receptor 358-372 Polyclonal antibody specifically detects Delta Opioid Receptor 358-372 in Mouse, Rat samples. It is validated for Immunocytochemistry, Immunofluorescence
Specifications
Specifications
Delta Opioid Receptor 358-372
Polyclonal
Unconjugated
OPRD1
Liquid
Rabbit
IgG
RUO
Antibody
Polyclonal
Mouse, Rat
Immunocytochemistry, Immunofluorescence
Conc. Not Determined
whole serum with 0.05% sodium azide
P32300, P33533
delta 1 opioid receptor; delta opioid receptor 1; Delta-type opioid receptor; DOR; D-OR-1; DOR-1; K56; mDOR; MSL-2; Nbor; Opioid receptor; Opioid receptor A; opioid receptor delta 1; opioid receptor, delta 1; OPRD; OPRD1; Ror-a
Human Delta Opioid Receptor 358-372
50 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
18386, 24613