Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Delta Opioid Receptor 358-372 Rabbit anti-Mouse, Rat, Polyclonal, Neuromics™ Non-distribution product as customer accommodation.

Rabbit Polyclonal Antibody

$187.24 - $448.88

Specifications

Antigen Delta Opioid Receptor 358-372
Concentration Conc. Not Determined
Applications Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-210-0021 Non-distribution product as customer accommodation.
View Documents
Neuromics, IncSupplier Diversity Partner
RA1010150UL
50 μL
Each for $187.24
Add to cart
 
50-210-0020 Non-distribution product as customer accommodation.
View Documents
Neuromics, IncSupplier Diversity Partner
RA10101150UL
150 μL
Each for $448.88
Add to cart
 
Description

Description

Delta Opioid Receptor 358-372 Polyclonal antibody specifically detects Delta Opioid Receptor 358-372 in Mouse, Rat samples. It is validated for Immunocytochemistry, Immunofluorescence
Specifications

Specifications

Delta Opioid Receptor 358-372
Immunocytochemistry, Immunofluorescence
Unconjugated
Rabbit
Mouse, Rat
P32300, P33533
18386, 24613
IgG
Primary
Antibody
Conc. Not Determined
Polyclonal
Liquid
RUO
whole serum with 0.05% sodium azide
delta 1 opioid receptor; delta opioid receptor 1; Delta-type opioid receptor; DOR; D-OR-1; DOR-1; K56; mDOR; MSL-2; Nbor; Opioid receptor; Opioid receptor A; opioid receptor delta 1; opioid receptor, delta 1; OPRD; OPRD1; Ror-a
Human Delta Opioid Receptor 358-372
Polyclonal
-20° C, Avoid Freeze/Thaw Cycles
OPRD1
SDS
Product Certifications
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit