Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

delta-Sarcoglycan Mouse anti-Human, Alexa Fluor™ 750, Clone: 19G8, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP242662AF750

Catalog No. NP242662A70


Add to cart

Description

Description

delta-Sarcoglycan Monoclonal antibody specifically detects delta-Sarcoglycan in Human samples. It is validated for Western Blot,ELISA.
Specifications

Specifications

delta-Sarcoglycan
19G8
Western Blot, ELISA
35 kDa dystrophin-associated glycoprotein, 35DAG, 35kD dystrophin-associated glycoprotein, CMD1LSG-delta, DAGD, delta-sarcoglycan, delta-SG, dystrophin associated glycoprotein, delta sarcoglycan, LGMD2F, MGC22567, placental delta sarcoglycan, sarcoglycan, delta (35kD dystrophin-associated glycoprotein), sarcoglycan, delta (35kDa dystrophin-associated glycoprotein), SGCDP, SGD
Mouse
IgG1b Kappa
Store at 4°C in the dark.
Monoclonal
Human
Western Blot, ELISA
Alexa Fluor 750
50 mM sodium borate with 0.05% sodium azide
SGCD
Recombinant human SGCD (57-289aa) purified from E. coli
100 μL
Primary
6444
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit