Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
delta-Sarcoglycan Antibody (19G8), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP242662F
Description
delta-Sarcoglycan Monoclonal specifically detects delta-Sarcoglycan in Human samples. It is validated for ELISA.Specifications
delta-Sarcoglycan | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
SGCD | |
Recombinant human SGCD (57-289aa) purified from E. coli | |
100 μL | |
Primary | |
Human | |
Purified |
ELISA | |
19G8 | |
ELISA | |
35 kDa dystrophin-associated glycoprotein, 35DAG, 35kD dystrophin-associated glycoprotein, CMD1LSG-delta, DAGD, delta-sarcoglycan, delta-SG, dystrophin associated glycoprotein, delta sarcoglycan, LGMD2F, MGC22567, placental delta sarcoglycan, sarcoglycan, delta (35kD dystrophin-associated glycoprotein), sarcoglycan, delta (35kDa dystrophin-associated glycoprotein), SGCDP, SGD | |
Mouse | |
Protein A purified | |
RUO | |
6444 | |
Store at 4C in the dark. | |
IgG1b κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction