Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ delta-Tetradecanolactone, 98%

Click to view available options
Quantity:
25g
100g
500g
2721-22-4
C14H26O2
226.36
MFCD00059809
SKQYTJLYRIFFCO-UHFFFAOYSA-N
delta-tetradecalactone,5-tetradecanolide,2h-pyran-2-one, tetrahydro-6-nonyl,delta-tetradecanolide,delta-tetradecanolactone,tetrahydro-6-nonyl-2h-pyran-2-one,fema no. 3590,6-nonyltetrahydro-2h-pyran-2-one,delta-myristolactone,5-hydroxytetradecanoic acid delta-lactone
520296
6-nonyloxan-2-one
CCCCCCCCCC1CCCC(=O)O1
2721-22-4
C14H26O2
226.36
MFCD00059809
SKQYTJLYRIFFCO-UHFFFAOYSA-N
delta-tetradecalactone,5-tetradecanolide,2h-pyran-2-one, tetrahydro-6-nonyl,delta-tetradecanolide,delta-tetradecanolactone,tetrahydro-6-nonyl-2h-pyran-2-one,fema no. 3590,6-nonyltetrahydro-2h-pyran-2-one,delta-myristolactone,5-hydroxytetradecanoic acid delta-lactone
520296
6-nonyloxan-2-one
CCCCCCCCCC1CCCC(=O)O1
Quantity 500g
Flash Point >110°C (230°F)
Melting Point 130°C to 135°C
Formula Weight 226.36
Percent Purity 98%
Chemical Name or Material delta-Tetradecanolactone

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA