Alfa Aesar™ delta-Valerolactam, 98+%

Manufacturer: Alfa Aesar™ A1227136

Quantity 500g

 View more versions of this product

Catalog No. AAA1227136

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 675-20-7
    Molecular Formula C5H9NO
    Formula Weight 99.13
    MDL Number MFCD00006037
    Beilstein 106434
    Synonym 2-Piperidone
    Chemical Name or Material delta-Valerolactam

    Specifications

    Formula Weight 99.13
    MDL Number MFCD00006037
    Synonym 2-Piperidone
    Melting Point 35° to 39°C
    Quantity 500g
    TSCA TSCA
    Molecular Formula C5H9NO
    CAS 675-20-7
    Beilstein 106434
    Flash Point 147°C (296°F)
    RTECS Number TO0110000
    Boiling Point 255° to 257°C
    Chemical Name or Material delta-Valerolactam
    Assay ≥98%

    Description

    Applications
    delta-Valerolactam is the starting material for a new route to hexahydrojulolidines, using intramolecular 1,4-cycloaddition. It is used as an intermediate in the preparation of other chemicals.

    Solubility
    Soluble in water.

    Notes
    Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit