Learn More
Selleck Chemical LLCSupplier Diversity Partner Deltarasin DFS Item
Manufacturer:  Selleck Chemical LLCSupplier Diversity Partner S72245MG
Deltarasin is a small molecular inhibitor of KRAS-PDEδ interaction with Kd of 38 nM for binding to purified PDEδ.
Catalog No. 50-194-8070