Learn More
Radians Deluxe HV Starter Kit
Meets various compliance standards and jobsite applications
Supplier: Radians RNHK8
Description
Includes:
- Foam earplugs (FP71)
- Corded with case
- Mirage safety eyewear (MR0110ID)
- Smoke frame/lens
- High visibility green cap style hard hat with 6 point ratchet suspension (GHR6-GREEN-HV)
- Glove high visibility knit glove textured rubber palm (RWG10)
- Large gloves
- Class 2 safety vest with zipper (SV2ZGMXL)
- X-large vest
Specifications
Deluxe HV Starter Kit | |
With bag |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Warning: Cancer and reproductive harm