Learn More
Deluxe Inclined Plane Accessories
SCI_ED
Manufacturer:  Lab Workx LLC 9036
 View more versions of this product
Catalog No. S40916
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Friction Block
  • Made of smooth finished maple or birch
  • With hook
  • Dimensions: 6.25 × 3 × 0.75 in.(15.9 × 7.6 × 1.9cm)
Friction Box
  • Open-top Baltic birch wooden box
  • Use to determine coefficient of static and kinetic friction
  • Features a hook at one end and two rubber strips secured to the bottom
  • Dimensions: 6.125 × 3.125 × 2.125 in. (15.6 × 7.9 × 5.4cm)
Specifications
Specifications
Friction Block
Deluxe inclined plane
Pulley